Learn More
Invitrogen™ TMEM30A Polyclonal Antibody

Description
Antibody detects endogenous levels of total CC50A.
Specifications
Specifications
| Antigen | TMEM30A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TMEM30A |
| Gene Accession No. | Q6AY41, Q8VEK0, Q9NV96 |
| Gene Alias | 2010200I23Rik; AW540225; C6orf67; Cdc50a; cell cycle control protein 50A; cell cycle control protein 50A; LOW QUALITY PROTEIN: cell cycle control protein 50A; Cell cycle control protein 50A-like protein; D9Wsu20e; FLJ10856; P4-ATPase flippase complex beta subunit TMEM30A; TMEM30A; Transmembrane protein 30A |
| Gene Symbols | TMEM30A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.