Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM30B Antibody, Novus Biologicals™
SDP

Catalog No. NBP15953420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15953420 20 μL
NBP159534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15953420 Supplier Novus Biologicals Supplier No. NBP15953420UL

Rabbit Polyclonal Antibody

TMEM30B Polyclonal specifically detects TMEM30B in Human samples. It is validated for Western Blot.

Specifications

Antigen TMEM30B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q3MIR4
Gene Alias cell cycle control protein 50B, transmembrane protein 30BCDC50BMGC126775
Gene Symbols TMEM30B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TMEM30B(transmembrane protein 30B) The peptide sequence was selected from the middle region of TMEM30B. Peptide sequence VYLYYELTNFYQNNRRYGVSRDDAQLSGLPSALRHPVNECAPYQRSAAGL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161291
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.