Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM49 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB001612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001612 0.025 mg
NB001592 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB001612 Supplier Novus Biologicals Supplier No. NBP278784SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM49 Polyclonal specifically detects TMEM49 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TMEM49
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 5 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 10 - 20 μg/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias DKFZp566I133, EPG3, TDC1, transmembrane protein 49TMEM49, vacuole membrane protein 1
Gene Symbols VMP1
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human TMEM49 (between amino acids 1-75) [UniProt Q96GC9]
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Autophagy, Cancer, ER Markers, Membrane Trafficking and Chaperones, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 81671
Target Species Human, Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.