Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM59L Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017248 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017248 Supplier Novus Biologicals Supplier No. NBP281937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM59L Polyclonal specifically detects TMEM59L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TMEM59L
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias BSMAPBrain-specific membrane-anchored protein, C19orf4, chromosome 19 open reading frame 4, transmembrane protein 59-like
Gene Symbols TMEM59L
Host Species Rabbit
Immunogen TMEM59L antibody was raised against a 19 amino acid synthetic peptide near carboxy terminus of human TMEM59L. The immunogen is located within amino acids 220 - 270 of TMEM59L. Amino Acid Squence: RVKTSSKAKVESEEPQDND
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25789
Test Specificity Multiple isoforms of TMEM59L are known to exist. TMEM59L antibody is predicted to not cross-react TMEM59.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.