Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM79 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15983220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15983220 20 μL
NBP159832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15983220 Supplier Novus Biologicals Supplier No. NBP15983220UL

Rabbit Polyclonal Antibody

TMEM79 Polyclonal specifically detects TMEM79 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TMEM79
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9BSE2
Gene Alias FLJ16057, FLJ32254, MGC13102, transmembrane protein 79
Gene Symbols TMEM79
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human TMEM79 (NP_115699). Peptide sequence LPLLSMLMWNLYYMFVVEPERMLTATESRLDYPDHARSASDYRPRPWG.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84283
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.