Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM79 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | TMEM79 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159832
![]() |
Novus Biologicals
NBP159832 |
100 μL |
Each for $499.50
|
|
|||||
NBP15983220
![]() |
Novus Biologicals
NBP15983220UL |
20 μL | N/A | N/A | N/A | ||||
Description
TMEM79 Polyclonal specifically detects TMEM79 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TMEM79 | |
Polyclonal | |
Rabbit | |
Q9BSE2 | |
84283 | |
Synthetic peptide directed towards the C terminal of human TMEM79 (NP_115699). Peptide sequence LPLLSMLMWNLYYMFVVEPERMLTATESRLDYPDHARSASDYRPRPWG. | |
Primary |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
FLJ16057, FLJ32254, MGC13102, transmembrane protein 79 | |
TMEM79 | |
IgG | |
43 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title