Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM87A Antibody (772807), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB373834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373834 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373834 Supplier Novus Biologicals Supplier No. FAB7966I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEM87A Monoclonal antibody specifically detects TMEM87A in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF

Specifications

Antigen TMEM87A
Applications Western Blot, Simple Western, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 772807
Conjugate Janelia Fluor 549
Dilution Western Blot, Simple Western, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DKFZp564G2022, PSEC0094, transmembrane protein 87A
Host Species Mouse
Immunogen E. coli-derived recombinant human TMEM87A, Ser22-Asn157, Accession # Q8NBN3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25963
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.