Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM97 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP130437A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP130437A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP130437A32 Supplier Novus Biologicals Supplier No. NBP130437AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM97 Polyclonal specifically detects TMEM97 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TMEM97
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MAC30Protein MAC30, transmembrane protein 97
Gene Symbols TMEM97
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-ProtNo. Q8VD00]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27346.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.