Learn More
Description
Specifications
Specifications
| Antigen | TMEM97 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | MAC30Protein MAC30, transmembrane protein 97 |
| Gene Symbols | TMEM97 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot: Q5BJF2] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
