Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEPAI, Mouse anti-Human, Clone: PMEPA1/2697, Novus Biologicals™
SDP

Catalog No. NB010531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010531 100 μg
NB010532 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010531 Supplier Novus Biologicals Supplier No. NBP279757100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen TMEPAI
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PMEPA1/2697
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein
Gene Symbols PMEPA1
Host Species Mouse
Immunogen Recombinant full-length human TMEPAI protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56937
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.