Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TMI 1, >98%
GREENER_CHOICESDP

Catalog No. 596050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-605-0 50 mg
59-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-605-0 Supplier Tocris Bioscience™ Supplier No. 5960/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adam 17 (TACE) and MMP inhibitor; orally bioavailable

Adam 17 (TACE) and MMP inhibitor (IC50 values are 3, 4.7, 6.6, 8.4, 12, 26 and 26 nM for MMP-13, MMP-2, MMP-1, ADAM 17, MMP-9, MMP-7 and MMP-14, respectively). Suppresses TNF-α production in an acute LPS-mouse model. Reduces severity score in an in vivo model of rheumatoid arthritis. Displays selective cytotoxicity to tumor cells and cancer stem cells in vitro. Induces tumor apoptosis in a breast cancer in vivo model. Orally bioavailable.

Chemical Identifiers

CAS 287403-39-8
Molecular Formula C17H22N2O5S2

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 398.49
Purity >98%
Molecular Weight (g/mol) 398.492
Chemical Name or Material (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.