Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TMI 1, >98%
GREENER_CHOICE

Catalog No. p-7121278
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Adam 17 (TACE) and MMP inhibitor; orally bioavailable

Adam 17 (TACE) and MMP inhibitor (IC50 values are 3, 4.7, 6.6, 8.4, 12, 26 and 26 nM for MMP-13, MMP-2, MMP-1, ADAM 17, MMP-9, MMP-7 and MMP-14, respectively). Suppresses TNF-α production in an acute LPS-mouse model. Reduces severity score in an in vivo model of rheumatoid arthritis. Displays selective cytotoxicity to tumor cells and cancer stem cells in vitro. Induces tumor apoptosis in a breast cancer in vivo model. Orally bioavailable.

Chemical Identifiers

CAS 287403-39-8
Molecular Formula C17H22N2O5S2

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 398.49
Purity >98%
Molecular Weight (g/mol) 398.492
Chemical Name or Material (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.