Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMIGD2 Antibody (953728), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346201 Supplier Novus Biologicals Supplier No. FAB8316MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen TMIGD2
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 953728
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line-derived human TMIGD2/CD28H, Met1-Gly150, Accession # Q96BF3
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126259
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.