Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMIGD2 Antibody (953730), Alexa Fluor™ 405, R&D Systems™
SDP

Catalog No. MAB83161AFB Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB83161AFB 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB83161AFB Supplier Novus Biologicals Supplier No. MAB83161AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TMIGD2
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 953730
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
Gene Symbols TMIGD2
Host Species Mouse
Immunogen Partial recombinant human CD28H protein (amino acids 23-150) [UniProt Q96BF3]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126259
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.