Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMIO, 98%, Thermo Scientific™

Catalog No. AC329350100
Click to view available options
Quantity:
10 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136440-22-7
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.159
MDL Number MFCD00168351
InChI Key CNIGTNPXWSLRPH-UHFFFAOYSA-N
Synonym tmio, 2,2,4-trimethyl-2h-imidazole 1-oxide, 2,2,4-trimethylimidazol-1-ium-1-olate, 2,2,4-trimethyl-2h-imidazole-1-oxide, 2,2,4-trimethyl-1-oxido-imidazol-1-ium, 2,2,4-trimethyl-2h-1.lambda.∼5∼-imidazol-1-ol, 2,2,4-trimethyl-2h-imidazole 1-oxide, for esr-spectroscopy
PubChem CID 379378
IUPAC Name 2,2,4-trimethyl-1-oxidoimidazol-1-ium
SMILES CC1=NC([N+](=C1)[O-])(C)C

Specifications

Melting Point 30°C to 40°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 10 mg
Formula Weight 126.2
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material TMIO, 98%
Hazard Category HYGROSCOPIC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.