Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMLHE Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB382841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382841 Supplier Novus Biologicals Supplier No. NBP306385IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMLHE Polyclonal antibody specifically detects TMLHE in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TMLHE
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BBOX2TML-alpha-ketoglutarate dioxygenase, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 2, EC 1.14.11.8, Epsilon-trimethyllysine 2-oxoglutarate dioxygenase, Epsilon-trimethyllysine hydroxylase, FLJ10727, TML dioxygenase, TMLD, TMLHTML hydroxylase, trimethyllysine dioxygenase, mitochondrial, trimethyllysine hydroxylase, epsilon, XAP130
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TMLHE fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 55217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.