Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMLHE Antibody, Novus Biologicals™
SDP

Catalog No. NBP255562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP255562 100 μL
NB435560 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP255562 Supplier Novus Biologicals Supplier No. NBP255562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMLHE Polyclonal specifically detects TMLHE in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TMLHE
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias BBOX2TML-alpha-ketoglutarate dioxygenase, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 2, EC 1.14.11.8, Epsilon-trimethyllysine 2-oxoglutarate dioxygenase, Epsilon-trimethyllysine hydroxylase, FLJ10727, TML dioxygenase, TMLD, TMLHTML hydroxylase, trimethyllysine dioxygenase, mitochondrial, trimethyllysine hydroxylase, epsilon, XAP130
Gene Symbols TMLHE
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IRYNNYDRAVINTVPYDVVHRWYTAHRTLTIELRRPENEFWVKLKPGRVLFIDNWRVLHGRECFTGYRQLCGCYLTRDDVLNTAR
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 55217
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.