Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tmp21/p23 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N110787A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N110787A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N110787A30 Supplier Novus Biologicals Supplier No. NB11057587AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Tmp21/p23
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein
Gene Symbols TMED10
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-ProtNo. P49755]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10972
Target Species Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus species
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.