Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tmp21/p23 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11057587T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11057587T 0.025 mL
NB11057587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11057587T Supplier Novus Biologicals Supplier No. NB11057587SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Tmp21/p23
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1 ug/ml, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P49755
Gene Alias 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein
Gene Symbols TMED10
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-Prot# P49755]
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10972
Target Species Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus species
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.