Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tmp21/p23 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Tmp21/p23
Concentration 0.95 mg/mL
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1 ug/ml, Immunohistochemistry-Paraffin, Knockdown Validated
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11057587T
SDP
View Documents
Novus Biologicals
NB11057587SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11057587
SDP
View Documents
Novus Biologicals
NB11057587
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Tmp21/p23 Polyclonal specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

Tmp21/p23
Western Blot 2 ug/ml, Immunohistochemistry 1 ug/ml, Immunohistochemistry-Paraffin, Knockdown Validated
Polyclonal
Rabbit
Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
PBS and 30% Glycerol with 0.1% Sodium Azide
21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein
TMED10
IgG
Affinity Purified
21 kDa
0.95 mg/mL
Western Blot, Immunohistochemistry
Unconjugated
RUO
Human, Mouse, Rat, Bovine, Primate, Rabbit, Xenopus species
P49755
10972
A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 50-150). [Swiss-Prot# P49755]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.