Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tmp21/p23 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB339249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339249 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339249 Supplier Novus Biologicals Supplier No. NB11057586PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tmp21/p23 Polyclonal antibody specifically detects Tmp21/p23 in Human, Mouse, Rat, Bovine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen Tmp21/p23
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS
Gene Alias 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human TMP21/TMED10 protein (within residues 100-200). [Swiss-Prot# P49755]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10972
Target Species Human, Mouse, Rat, Bovine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.