Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS11A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241140 0.1 mg
NB396486 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241140 Supplier Novus Biologicals Supplier No. NBP241140
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

TMPRSS11A Polyclonal specifically detects TMPRSS11A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TMPRSS11A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA
Formulation PBS with 0.02% Sodium Azide
Gene Alias Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11A, Transmembrane Protease, Serine 11A
Gene Symbols TMPRSS11A
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG1. The immunogen is located within amino acids 300 - 350 of ECRG1. Amino Acid Squence: YGGESQNDLREARVKI
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339967
Test Specificity At least two isoforms of ECRG1 are known to exist; ECRG1 antibody is predicted to not cross-react with other ECRG family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.