Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS11A Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343990 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343990 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343990 Supplier Novus Biologicals Supplier No. NBP241140MFV500

Chicken Polyclonal Antibody

TMPRSS11A Polyclonal antibody specifically detects TMPRSS11A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen TMPRSS11A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11A, Transmembrane Protease, Serine 11A
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG1. The immunogen is located within amino acids 300 - 350 of ECRG1. Amino Acid Squence: YGGESQNDLREARVKI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 339967
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.