Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMPRSS11A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
| Antigen | TMPRSS11A |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TMPRSS11A Polyclonal antibody specifically detects TMPRSS11A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TMPRSS11A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| PBS, pH 7.2, 40% glycerol | |
| 339967 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11A, Transmembrane Protease, Serine 11A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FIDSAWKKNYIKNQVVRLTPEEDGVKVDVIMVFQFPSTEQRAVREKKIQSILNQKIR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title