Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS2 Antibody (1038105), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339919 Supplier Novus Biologicals Supplier No. FAB107231MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMPRSS2 Monoclonal specifically detects TMPRSS2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TMPRSS2
Applications Flow Cytometry
Classification Monoclonal
Clone 1038105
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2
Host Species Mouse
Immunogen Human TMPRSS2 synthetic peptide, Accession # O15393
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7113
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.