Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB050190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050190 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050190 Supplier Novus Biologicals Supplier No. NBP297969AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMPRSS2 Polyclonal specifically detects TMPRSS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TMPRSS2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2
Host Species Rabbit
Immunogen TMPRSS2 Antibody was produced in rabbits immunized with E. coli-derived human TMPRSS2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7113
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.