Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS4 Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB050227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050227 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050227 Supplier Novus Biologicals Supplier No. NBP297970R

Rabbit Polyclonal Antibody

TMPRSS4 Polyclonal specifically detects TMPRSS4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TMPRSS4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CAPH2, Channel-activating protease 2, EC 3.4.21, EC 3.4.21.4, EC 3.4.21.43, Membrane-type serine protease 2, MT-SP2, TMPRSS3EC 3.4.21.-, transmembrane protease serine 4, transmembrane protease, serine 4, transmembrane serine protease 3, Type II membrane serine protease
Host Species Rabbit
Immunogen TMPRSS4 Antibody is produced in rabbits immunized with E. coli-derived Human TMPRSS4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Developmental Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 56649
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.