Learn More
Description
Specifications
Specifications
| Antigen | TMTC4 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.2-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | FLJ14624, FLJ22153, transmembrane and tetratricopeptide repeat containing 4, transmembrane and TPR repeat-containing protein 4 |
| Gene Symbols | TMTC4 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to TMTC4(transmembrane and tetratricopeptide repeat containing 4) The peptide sequence was selected from the N terminal of TMTC4. Peptide sequence NKDLQAETPLGDLWHHDFWGSRLSSNTSHKSYRPLTVLTFRINYYLSGGF. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
