Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMTC4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | TMTC4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15992920
![]() |
Novus Biologicals
NBP15992920UL |
20 μL |
Each for $158.00
|
|
|||||
NBP159929
![]() |
Novus Biologicals
NBP159929 |
100 μL |
Each for $487.50
|
|
|||||
Description
TMTC4 Polyclonal specifically detects TMTC4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMTC4 | |
Polyclonal | |
Rabbit | |
FLJ14624, FLJ22153, transmembrane and tetratricopeptide repeat containing 4, transmembrane and TPR repeat-containing protein 4 | |
TMTC4 | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
84899 | |
Synthetic peptides corresponding to TMTC4(transmembrane and tetratricopeptide repeat containing 4) The peptide sequence was selected from the N terminal of TMTC4. Peptide sequence NKDLQAETPLGDLWHHDFWGSRLSSNTSHKSYRPLTVLTFRINYYLSGGF. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title