Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMUB2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB329461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329461 Supplier Novus Biologicals Supplier No. NBP300175PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMUB2 Polyclonal antibody specifically detects TMUB2 in Human samples. It is validated for ELISA

Specifications

Antigen TMUB2
Applications ELISA
Classification Polyclonal
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias MGC3123, transmembrane and ubiquitin-like domain containing 2, transmembrane and ubiquitin-like domain-containing protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human TMUB2 (Accession#: NP_803190.2; Tyr37-Ser245)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79089
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.