Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tn mIgM (STn-PS A1), clone KT-IgM-8 Mouse anti-Human, Unconjugated, Clone: Kt-IgM-8, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $481.52
Specifications
Antigen | Tn mIgM (STn-PS A1), clone KT-IgM-8 |
---|---|
Clone | Kt-IgM-8 |
Applications | ELISA, Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC17172MI
|
MilliporeSigma
MABC171725UL |
25 μL |
Each for $189.29
|
|
|||||
MABC17171MI
|
MilliporeSigma
MABC1717100UL |
100 μL |
Each for $481.52
|
|
|||||
Description
Tn mIgM (STn-PS A1), clone KT-IgM-8 Monoclonal antibody specifically detects Tn mIgM (STn-PS A1), clone KT-IgM-8 in Human samples. It is validated for ELISA, Flow Cytometry, Function Assay.Specifications
Tn mIgM (STn-PS A1), clone KT-IgM-8 | |
ELISA, Flow Cytometry, Functional Assay | |
Unconjugated | |
Mouse | |
Human | |
Semi-synthetic Tn-PS A1 (Tn= D-GalpNAc) | |
Primary | |
Stable for 1 year at -20°C from date of receipt. |
Kt-IgM-8 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
Thomsen-nouveau (Tn);Tn antigen;Tn saccharide;N-acetyl Galactosamine;Alpha-GalNAc;Alpha GalpNAc;GalNAc;GalNAc-alpha;N-acetyl-alpha-D-galactosamine;Blood group A antigen | |
IgM | |
Ligatrap | |
Clone KT-IgM-8 is a mouse monoclonal antibody that detects Thomsen-nouveau (Tn) polysaccharide A1 monovalently, including in human tumor cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title