Learn More
TNF-alpha Antibody (CDP 571 (Humicade)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | TNF-alpha |
| Applications | ELISA, Immunohistochemistry, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | CDP 571 (Humicade) |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000 |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Gene Alias | APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Gene Symbols | TNF |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.