Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Antibody (CDP 571 (Humicade)) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$268.00 - $584.50

Specifications

Antigen TNF-alpha
Clone CDP 571 (Humicade)
Dilution ELISA 1:100 - 1:2000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25267925
SDP
View Documents
Novus Biologicals
NBP2526790.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP25267902
SDP
View Documents
Novus Biologicals
NBP2526790.2MG
0.2 mg
Each for $584.50
Only null left
Add to Cart
 
Description

Description

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization.
Specifications

Specifications

TNF-alpha
ELISA 1:100 - 1:2000
Monoclonal
Purified
RUO
Human
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
TNF
IgG
Protein A or G purified
The antibody binds to human TNF-alpha with a Kd of 0.087 nM.
CDP 571 (Humicade)
ELISA
Unconjugated
Rabbit
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
7124
Human TNF-alpha
Primary
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.