Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Antibody (D2E7 (Adalimumab)), PerCP, Novus Biologicals™
SDP

Catalog No. NB343054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343054 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343054 Supplier Novus Biologicals Supplier No. NBP262568PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Neutralization

Specifications

Antigen TNF-alpha
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Neutralization
Classification Monoclonal
Clone D2E7 (Adalimumab)
Conjugate PerCP
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Block/Neutralize
Formulation PBS
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Host Species Rabbit
Immunogen This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha by guided phage-display technology, using the murine anti-hTNF alpha antibody Mab32 as a template (c.f. also US Patent 6090382).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7124
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.