Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TNF-alpha Antibody Pair [HRP]
SDP

Catalog No. p-7183134 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010389 15 Plates
NB010390 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010389 Supplier Novus Biologicals™ Supplier No. NBP27967515PLATES
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TNF-alpha.

Specifications

Assay Range 23.44 pg/mL
Assay Sensitivity 12.5-800 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TNF-alpha.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene ID (Entrez) 7124
Gene Symbol TNF
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human TNF-alpha monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human TNF-alpha monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 38 ng of recombinant Human TNF-alpha.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
    Storage Requirements Storage is content dependent.
    Target TNF-alpha
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1.2 μg/mL in Dilution Buffer before use. 3. Standard: Reconstitute with 1 ml Dilution Buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 800 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.