Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Antibody (TNF706 + P/T2) - IHC-Prediluted, Novus Biologicals™
SDP

Catalog No. NBP248471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248471 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP248471 Supplier Novus Biologicals Supplier No. NBP248471
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human, Mouse, Rat, Canine, Feline, Rabbit, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TNF-alpha
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TNF706 + P/T2
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen Recombinant N-terminal fragment of human TNF-alpha (TNF706); Peptide corresponding to aa115-130 of human TNF-alpha (P/T2) (Uniprot: P01375)
Molecular Weight of Antigen 17 kDa
Purification Method Protein L or PEG purified
Quantity 7 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity TNF alpha is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17kDa band following SDS PAGE under non-reducing conditions. TNF alpha causes cytolysis of certain transformed cells. Although it has little effect on many cultured normal human cells, TNF alpha appears to be directly toxic to vascular endothelial cells. Other actions of TNF alpha include stimulating growth of human fibroblasts and other cell lines, activating polymorphonuclear neutrophils and osteoclasts, and induction of interleukin 1, prostaglandin E2 and collagenase production. TNF alpha is currently being evaluated in treatment of certain cancers and AIDS Related Complex.
Target Species Human, Mouse, Rat, Canine, Feline, Rabbit, Zebrafish
Content And Storage Store at 4C.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.