Learn More
TNF Mouse anti-Human, PE, Clone: MAB11, BD
Mouse Monoclonal Antibody
$122.00 - $284.00
Specifications
Antigen | TNF |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Host Species | Mouse |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB562083
|
BD Biosciences
562083 |
25 Tests |
Each for $122.00
|
|
|||||
BDB559321
|
BD Biosciences
559321 |
100 Tests |
Each for $284.00
|
|
|||||
Description
The MAb11 monoclonal antibody specifically binds to human tumor necrosis factor (TNF, also known as TNF- α ) protein. TNF is an efficient juxtacrine, paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN- γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The MAb11 antibody has been reported to crossreact with Rhesus Macaque TNF.
Specifications
TNF | |
Monoclonal | |
Mouse | |
Human | |
Recombinant Human TNF | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.