Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RI/TNFRSF1A Antibody (001), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB108795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108795 Supplier Novus Biologicals Supplier No. NBP289687AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TNF RI/TNFRSF1A
Applications Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TNF RI/TNFRSF1A (NP_001056.1; Met1-Thr211).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7132
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.