Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RI/TNFRSF1A Antibody (028), DyLight 350
SDP

Catalog No. NB075627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075627 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075627 Supplier Novus Biologicals Supplier No. NBP290613UV

Rabbit Monoclonal Antibody

TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen TNF RI/TNFRSF1A
Applications ELISA
Classification Monoclonal
Clone 28
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNF RI/TNFRSF1A (NP_035739.2; Met 1-Ala 212).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7132
Test Specificity No cross-reactivity in ELISA with: Human TNFRII
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.