Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RI/TNFRSF1A Antibody (RM0011-6F13), Alexa Fluor™ 750
SDP

Catalog No. N1108469A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1108469A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1108469A70 Supplier Novus Biologicals Supplier No. NB11085469AF750

Armenian Hamster Monoclonal Antibody

TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen TNF RI/TNFRSF1A
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone RM0011-6F13
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Gene Symbols TNFRSF1A
Host Species Armenian Hamster
Immunogen Mouse TNF RI/TNFRSF1A (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7132.0
Test Specificity This is specific to the extracellular domain of mouse TNFR-1 protein.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.