Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RII/TNFRSF1B Antibody (RM0012-6F13) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018038 0.025 mg
NB11085470 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018038 Supplier Novus Biologicals Supplier No. NB110854700.025MG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen TNF RII/TNFRSF1B
Applications CyTOF, Flow Cytometry, ELISA, Western Blot, Immunoprecipitation
Classification Monoclonal
Clone RM0012-6F13
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:100-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:100-1:200, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Gene Symbols TNFRSF1B
Host Species Armenian Hamster
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from an Armenian Hamster immunized with mouse Recombinant extracellular domain of mouse TNFR-II protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7133
Test Specificity This is specific to the extracellular domain of mouse TNFR-II protein.
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.