Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP8 Antibody (01), PE, Novus Biologicals™
SDP

Catalog No. NB351216 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351216 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351216 Supplier Novus Biologicals Supplier No. NBP306584PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TNFAIP8 Monoclonal antibody specifically detects TNFAIP8 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TNFAIP8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 01
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias GG2-1, Head and neck tumor and metastasis-related protein, MDC-3.13TNF-induced protein GG2-1, NDED, NF-kappa-B-inducible DED-containing protein, SCC-S2SCCS2, TNF alpha-induced protein 8, tumor necrosis factor alpha-induced protein 8, tumor necrosis factor, alpha-induced protein 8
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFAIP8 (Uniprot#: O95379; Met1-Ile198).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 25816
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.