Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP8 Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB087268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087268 Supplier Novus Biologicals Supplier No. NBP300317IR

Rabbit Polyclonal Antibody

TNFAIP8 Polyclonal specifically detects TNFAIP8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TNFAIP8
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias GG2-1, Head and neck tumor and metastasis-related protein, MDC-3.13TNF-induced protein GG2-1, NDED, NF-kappa-B-inducible DED-containing protein, SCC-S2SCCS2, TNF alpha-induced protein 8, tumor necrosis factor alpha-induced protein 8, tumor necrosis factor, alpha-induced protein 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant TNFAIP8 (O95379; Met1-Ile198).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 25816
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.