Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRH3/TNFRSF26 Antibody (509027), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312064 Supplier Novus Biologicals Supplier No. FAB5330APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TNFRH3/TNFRSF26 Monoclonal antibody specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen TNFRH3/TNFRSF26
Applications Flow Cytometry
Classification Monoclonal
Clone 509027
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias APO-3;DDR3;DR3;GEF720;LARD;PLEKHG5;TNFRSF12;TNFRSF25;TR3;TRAMP;Tumor necrosis factor receptor superfamily member 25;WSL-1;WSL-LR
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26, Ser20-Lys162, Accession # P83626
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.