Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNIK Antibody (HL1751) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294892 Supplier Novus Biologicals Supplier No. NBP325725
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TNIK Monoclonal antibody specifically detects TNIK in Human samples. It is validated for Western Blot

Specifications

Antigen TNIK
Applications Western Blot
Classification Monoclonal
Clone HL1751
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS
Gene Alias EC 2.7.11, KIAA0551EC 2.7.11.1, TRAF2 and NCK interacting kinase, TRAF2 and NCK-interacting protein kinase
Host Species Rabbit
Immunogen Synthetic peptide of human TNIK.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23043
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.