Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNKS2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220675 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220675 Supplier Novus Biologicals Supplier No. NBP220675

Rabbit Polyclonal Antibody

TNKS2 Polyclonal specifically detects TNKS2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TNKS2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.25mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9H2K2
Gene Alias EC 2.4.2.30, PARP-5b, PARP5BTankyrase-like protein, PARP-5c, PARP5C, pART6, Poly [ADP-ribose] polymerase 5B, TANK2Tankyrase-related protein, tankyrase 2, Tankyrase II, tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2, tankyrase-2, TNKLTNKS-2, TRF1-interacting ankyrin-related ADP-ribose polymerase 2
Gene Symbols TNKS2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Tankyrase 2. The exact sequence is proprietary.
Molecular Weight of Antigen 127 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80351
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Rat (92%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.