Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNKS2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP136993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP136993 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP136993 Supplier Novus Biologicals Supplier No. NBP136993
Add to Cart
Add to Cart

Goat Polyclonal Antibody

TNKS2 Polyclonal specifically detects TNKS2 in Human samples. It is validated for Western Blot.

Specifications

Antigen TNKS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_079511.1
Gene Alias EC 2.4.2.30, PARP-5b, PARP5BTankyrase-like protein, PARP-5c, PARP5C, pART6, Poly [ADP-ribose] polymerase 5B, TANK2Tankyrase-related protein, tankyrase 2, Tankyrase II, tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2, tankyrase-2, TNKLTNKS-2, TRF1-interacting ankyrin-related ADP-ribose polymerase 2
Gene Symbols TNKS2
Host Species Goat
Immunogen Peptide with sequence RECOMBINANT2 corresponding to internal region according to NP_079511.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80351
Test Specificity Fusion protein containing 73 aas from region aa800-900 (according to NP_079511.1). No crossreactivity expected with TANK1.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.