Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNNC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 135041AP150UL
Description
Troponin, working in conjunction with tropomyosin, functions as a molecular switch, regulating muscle contraction in response to changes in the intracellular Ca2+ concentration. Troponin consists of three subunits: the Ca2+-binding subunit troponin C (TnC), the tropomyosin-binding subunit troponin T (TnT), and the inhibitory subunit troponin I (TnI). In response to beta-adrenergic stimulation of the heart, Ser23 and Ser24 of TnI (cardiac) are phosphorylated by PKA and PKC. This phosphorylation stimulates a conformational change of the regulatory domain of TnC, reduces the association between TnI and TnC, and decreases myofilament Ca2+ sensitivity by reducing the Ca2+ binding affinity of TnC (1-3).Specifications
| TNNC1 | |
| Polyclonal | |
| Unconjugated | |
| TNNC1 | |
| CMD1Z, TN C, TNC, TNNC, TNNC1, troponin C type 1 (slow) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21924, 290561, 7134 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19123, P63316 | |
| Tnnc1 | |
| TNNC1 Fusion Protein Ag4339 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction