Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNNI3K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TNNI3K |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17927920
![]() |
Novus Biologicals
NBP17927920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179279
![]() |
Novus Biologicals
NBP179279 |
100 μL |
Each for $487.50
|
|
|||||
Description
TNNI3K Polyclonal specifically detects TNNI3K in Mouse samples. It is validated for Western Blot.Specifications
TNNI3K | |
Western Blot | |
Unconjugated | |
RUO | |
CARKCardiac ankyrin repeat kinase, EC 2.7.11.1, MGC142099, MGC33828, serine/threonine-protein kinase TNNI3K, TNNI3 interacting kinase, TNNI3-interacting kinase | |
TNNI3K | |
IgG |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
NP_796040 | |
51086 | |
The immunogen for this antibody is Tnni3k. Peptide sequence NLVACDPSRSSGEKDEQTCLMWAYEKGHDAIVTLLKHYKRPQDELPCNEY. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title