Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNP, Tocris Bioscience™

Catalog No. p-4816808
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of IP6K; also inhibits IP3K

Reversible inositol hexakisphosphate kinase (IP6K) inhibitor (IC50 = 0.47 μM for inhibition of InsP7 formation). Also inhibits inositol 1,4,5-trisphosphate 3-kinase (IP3K) (IC50 = 10.2 μM). Binds to the ATP binding site of IP3K (Ki = 4.3 μM).

Chemical Identifiers

CAS 519178-28-0
Molecular Formula C20H16F3N7O2
Molecular Weight (g/mol) 443.39
InChI Key DDSBPUYZPWNNGH-UHFFFAOYSA-N
Synonym ip3k inhibitor, tnp, inositol-1,4,5-trisphosphate 3-kinase inhibitor, n2-m-trifluorobenzyl , n6-p-nitrobenzyl purine, n6-4-nitrophenyl methyl-n2-3-trifluoromethyl phenyl methyl-9h-purine-2,6-diamine, purine, 5, tnp hplc, n2-3-trifluoromethyl benzyl-n6-4-nitrobenzyl-9h-purine-2,6-diamine, n∼6∼-4-nitrophenyl methyl-n∼2∼-3-trifluoromethyl phenyl methyl-7h-purine-2,6-diamine
PubChem CID 16760513
IUPAC Name 6-N-[(4-nitrophenyl)methyl]-2-N-[[3-(trifluoromethyl)phenyl]methyl]-7H-purine-2,6-diamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CNC2=NC3=C(C(=N2)NCC4=CC=C(C=C4)[N+](=O)[O-])NC=N3

Specifications

Quantity 10 mg
Formula Weight 443.38
Percent Purity >97%
Chemical Name or Material TNP
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.